Learn More
MSC SPI Height Gage Accessories, Spare Part Kit, Double Beam Electronic Height Gages, Scriber Clamp
Catalog No. 5000529074
Encompass
This spare part kit provides essential accessories for double beam electronic height gages, including a scriber clamp, indicator bar and clamp, and a Z shape scriber. It is designed to support the functionality of your height gage operations.
  • Spare part kit.
  • For use with double beam electronic height gages.
  • Includes scriber clamp.
  • Includes indicator bar and clamp.
  • Includes Z shape scriber.
Catalog No. 50-005-29074 Supplier MSC Supplier No. 39387543
This spare part kit provides essential accessories for double beam electronic height gages, including a scriber clamp, indicator bar and clamp, and a Z shape scriber. It is designed to support the functionality of your height gage operations.
  • Spare part kit.
  • For use with double beam electronic height gages.
  • Includes scriber clamp.
  • Includes indicator bar and clamp.
  • Includes Z shape scriber.