Learn More
MSC SPI Horizontal Dial Test Indicator, 0.0300 in Max, 0-100-0 Dial, 0.0005 in Accuracy
Catalog No. 5000532284
Encompass
This Horizontal Dial Test Indicator is designed for measuring geometric tolerances and dimensional deviations of workpieces, especially in confined spaces. It offers a maximum measurement of 0.0300 inches, a 0-100-0 dial reading, and an accuracy of 0.000500 inches. The indicator features a 1-1/4 inch black dial with 0.000500 inch graduation.
  • Fully jeweled movement
  • Satin chrome finish
  • Universal positioning capability
  • Easy-to-read dial
  • Supplied with 3/8 in dovetail stems
  • Short form calibration certificate included
  • Furnished in fitted case
  • Two year limited warranty
Catalog No. 50-005-32284 Supplier MSC Supplier No. 37543725
This Horizontal Dial Test Indicator is designed for measuring geometric tolerances and dimensional deviations of workpieces, especially in confined spaces. It offers a maximum measurement of 0.0300 inches, a 0-100-0 dial reading, and an accuracy of 0.000500 inches. The indicator features a 1-1/4 inch black dial with 0.000500 inch graduation.
  • Fully jeweled movement
  • Satin chrome finish
  • Universal positioning capability
  • Easy-to-read dial
  • Supplied with 3/8 in dovetail stems
  • Short form calibration certificate included
  • Furnished in fitted case
  • Two year limited warranty