Learn More
MSC SPI Indicator Contact Point Extension, 4 in Length, #4-48 Thread, Steel
Catalog No. 5000512354
Encompass
The SPI Indicator Contact Point Extension is designed for use with AGD dial indicators and any test indicator on your dial indicator. It features a #4-48 thread and is constructed from durable steel, offering a 4-inch extension for various measurement applications.
  • Compatible with AGD dial indicators
  • Constructed from steel
  • Features a #4-48 thread size
  • Suitable for any test indicator on your dial indicator
  • Has an overall diameter of 7/32 inch
  • Equipped with female #4-48 on one end and male #4-48 on the other
Catalog No. 50-005-12354 Supplier MSC Supplier No. 35915545
The SPI Indicator Contact Point Extension is designed for use with AGD dial indicators and any test indicator on your dial indicator. It features a #4-48 thread and is constructed from durable steel, offering a 4-inch extension for various measurement applications.
  • Compatible with AGD dial indicators
  • Constructed from steel
  • Features a #4-48 thread size
  • Suitable for any test indicator on your dial indicator
  • Has an overall diameter of 7/32 inch
  • Equipped with female #4-48 on one end and male #4-48 on the other