Learn More
MSC SPI Indicator Positioner & Holder, Fine Adjustment, Articulated Arm, 285 lb Magnetic Pull
Catalog No. 5000530448
Encompass
The SPI Indicator Positioner & Holder is an articulated arm device designed for precise positioning of indicators. It features fine adjustment capabilities and includes a magnetic base with a strong 285 lb pull for secure mounting.
  • Flexible adjustment from multiple angles
  • Strong and controllable adsorption force
  • Simple and quick to operate
Catalog No. 50-005-30448 Supplier MSC Supplier No. 94110053
The SPI Indicator Positioner & Holder is an articulated arm device designed for precise positioning of indicators. It features fine adjustment capabilities and includes a magnetic base with a strong 285 lb pull for secure mounting.
  • Flexible adjustment from multiple angles
  • Strong and controllable adsorption force
  • Simple and quick to operate