Learn More
MSC SPI Indicator Positioner & Holder, Fine Adjustment, Articulated Arm, 310 mm Post Height
Catalog No. 5000530449
Encompass
The Indicator Positioner and Holder features an articulated arm and fine adjustment, providing flexible positioning from multiple angles. Constructed from durable aluminum and steel, this tool is suitable for a variety of laboratory and industrial applications requiring precise indicator placement.
  • Articulated arm for flexible positioning
  • Fine adjustment for precise placement
  • Constructed from aluminum and steel
  • Magnetic base type
  • M8 internal thread base mount type
  • Post height of 310 mm
Catalog No. 50-005-30449 Supplier MSC Supplier No. 94110079
The Indicator Positioner and Holder features an articulated arm and fine adjustment, providing flexible positioning from multiple angles. Constructed from durable aluminum and steel, this tool is suitable for a variety of laboratory and industrial applications requiring precise indicator placement.
  • Articulated arm for flexible positioning
  • Fine adjustment for precise placement
  • Constructed from aluminum and steel
  • Magnetic base type
  • M8 internal thread base mount type
  • Post height of 310 mm