Learn More
MSC SPI Magnetic Surface Gage and Indicator Set, 0.0005 in. Graduation, 0-15-0 Dial Reading, 1 ea
Catalog No. 5000513575
Encompass
This magnetic surface gage and dial test indicator kit is a versatile tool designed for precision measurement and layout applications. The set includes a high-quality dial test indicator and a heavy-duty magnetic base featuring an integrated on/off switch and fine adjustment capability, allowing for secure and stable positioning during use.
  • Includes dial test indicator with 1 in. dial diameter and 0-15-0 reading
  • Features 0.0005 in. graduation
  • Magnetic base measures 3-1/8 in. x 2-7/16 in. x 1-7/16 in.
  • Includes two dovetail stems (5/32 in. and 3/8 in. diameters)
  • Equipped with two holding rods and a 7/32 in. x 4 in. scriber rod
  • Doubles as a surface gage when the magnet is switched off
  • Supplied with a traceable short form certificate of calibration to NIST
Catalog No. 50-005-13575 Supplier MSC Supplier No. 01579408
This magnetic surface gage and dial test indicator kit is a versatile tool designed for precision measurement and layout applications. The set includes a high-quality dial test indicator and a heavy-duty magnetic base featuring an integrated on/off switch and fine adjustment capability, allowing for secure and stable positioning during use.
  • Includes dial test indicator with 1 in. dial diameter and 0-15-0 reading
  • Features 0.0005 in. graduation
  • Magnetic base measures 3-1/8 in. x 2-7/16 in. x 1-7/16 in.
  • Includes two dovetail stems (5/32 in. and 3/8 in. diameters)
  • Equipped with two holding rods and a 7/32 in. x 4 in. scriber rod
  • Doubles as a surface gage when the magnet is switched off
  • Supplied with a traceable short form certificate of calibration to NIST