Learn More
MSC SPI Individual Rectangular Gage Block, 0.30000 in, Grade AS-1, Steel
Catalog No. 5000510256
Encompass
This individual rectangular steel gage block is designed for precision measurement applications. Manufactured to Grade AS-1, it ensures accuracy and durability for critical tasks. Each block is engineered with features that enhance its performance and longevity in demanding environments.
  • Each gage block has blended radii and is burr-proofed.
  • Wringing or stacking the block will not cause damage to the measuring surfaces.
  • Certificate of calibration traceable to NIST included with each gage.
Catalog No. 50-005-10256 Supplier MSC Supplier No. 87877320
This individual rectangular steel gage block is designed for precision measurement applications. Manufactured to Grade AS-1, it ensures accuracy and durability for critical tasks. Each block is engineered with features that enhance its performance and longevity in demanding environments.
  • Each gage block has blended radii and is burr-proofed.
  • Wringing or stacking the block will not cause damage to the measuring surfaces.
  • Certificate of calibration traceable to NIST included with each gage.