Learn More
MSC SPI Inspection & Quality Label, 'Calibrate Before Each Use', 2 inch, Removable Vinyl
Catalog No. 5000511948
Encompass
This Inspection & Quality Label is designed for clear identification of equipment that requires calibration. Featuring the message 'Calibrate Before Each Use', this 2-inch wide vinyl label comes with a removable adhesive, ensuring easy application and removal without residue. Each pack contains 160 labels.
  • Label message: Calibrate before each use
  • Overall width: 2 inch
  • Material: Vinyl
  • Adhesion type: Removable
  • Comes in packs of 160
  • Features a write-on surface
  • Red and white color scheme
  • Rectangle shape for clear display
Catalog No. 50-005-11948 Supplier MSC Supplier No. 71203830
This Inspection & Quality Label is designed for clear identification of equipment that requires calibration. Featuring the message 'Calibrate Before Each Use', this 2-inch wide vinyl label comes with a removable adhesive, ensuring easy application and removal without residue. Each pack contains 160 labels.
  • Label message: Calibrate before each use
  • Overall width: 2 inch
  • Material: Vinyl
  • Adhesion type: Removable
  • Comes in packs of 160
  • Features a write-on surface
  • Red and white color scheme
  • Rectangle shape for clear display