Learn More
MSC SPI Inspection Surface Plate Stand, Stationary, Steel, 36 in Plate Length, 36 in Height
Catalog No. 5000513190
Encompass
This inspection surface plate stand is a stationary, steel unit with a 36 inch plate length and a 36 inch height, designed for use with precision granite surface plates. It provides a stable and reliable platform for various inspection and measurement applications.
  • Stationary design
  • Durable steel construction
  • Compatible with precision granite surface plates
  • Includes two accuracy certificates
  • Complies with Fed Spec GGG-P-463c, ISO 9001, ASME B89.3.7-2013, ISO 17025 standards
  • NIST traceable certificated
  • Gray color
  • Manufactured in the United States
Catalog No. 50-005-13190 Supplier MSC Supplier No. 01940568
This inspection surface plate stand is a stationary, steel unit with a 36 inch plate length and a 36 inch height, designed for use with precision granite surface plates. It provides a stable and reliable platform for various inspection and measurement applications.
  • Stationary design
  • Durable steel construction
  • Compatible with precision granite surface plates
  • Includes two accuracy certificates
  • Complies with Fed Spec GGG-P-463c, ISO 9001, ASME B89.3.7-2013, ISO 17025 standards
  • NIST traceable certificated
  • Gray color
  • Manufactured in the United States