Learn More
MSC SPI Interchangeable Rod Micrometer, 1 to 2 Inch, Mechanical, 0.00035 in Accuracy
Catalog No. 5000530277
Encompass
This mechanical interchangeable rod micrometer provides precise measurements within a 1 to 2 inch range. It features a 0.001 inch graduation and an accuracy of 0.00035 inch, making it suitable for various demanding applications.
  • Each rod is marked for range
  • 0.001 inch graduation
  • Accurate up to 0.0004 inch
  • 0.248 inch rod diameter
  • 1/4 inch travel
  • Includes 2 rods
  • Includes NIST traceability certification
  • Includes short form certificate of calibration
  • Includes case
Catalog No. 50-005-30277 Supplier MSC Supplier No. 76831700
This mechanical interchangeable rod micrometer provides precise measurements within a 1 to 2 inch range. It features a 0.001 inch graduation and an accuracy of 0.00035 inch, making it suitable for various demanding applications.
  • Each rod is marked for range
  • 0.001 inch graduation
  • Accurate up to 0.0004 inch
  • 0.248 inch rod diameter
  • 1/4 inch travel
  • Includes 2 rods
  • Includes NIST traceability certification
  • Includes short form certificate of calibration
  • Includes case