Learn More
MSC SPI Loupe, 15x Magnification, Triplet Lens, 21.03 mm
Catalog No. 5000531596
Encompass
This loupe features lenses made of three precision ground elements, cemented together to provide a clear and sharp image for detailed inspection.
  • Triplet lens type
  • 15x magnification
  • 21.03 mm lens diameter
  • Hand-held design
  • Lenses constructed from 3 precision ground elements
  • Foldable
  • No light source included
Catalog No. 50-005-31596 Supplier MSC Supplier No. 88441134
This loupe features lenses made of three precision ground elements, cemented together to provide a clear and sharp image for detailed inspection.
  • Triplet lens type
  • 15x magnification
  • 21.03 mm lens diameter
  • Hand-held design
  • Lenses constructed from 3 precision ground elements
  • Foldable
  • No light source included