Learn More
MSC SPI Loupe, 20x Magnification, 53/64 in Lens Diameter, Triplet Lens
Catalog No. 5000534549
Encompass
This loupe provides 20x magnification with a triplet lens design for clear, detailed viewing. It is a hand-held magnifier featuring three precision ground lens elements, making it suitable for various inspection tasks.
  • Triplet lens type
  • 20x minimum magnification
  • 53/64 in (21.03 mm) lens diameter
  • Hand-held design
  • Features 3 precision ground lens elements
  • Foldable design
  • No light source included
Catalog No. 50-005-34549 Supplier MSC Supplier No. 88441142
This loupe provides 20x magnification with a triplet lens design for clear, detailed viewing. It is a hand-held magnifier featuring three precision ground lens elements, making it suitable for various inspection tasks.
  • Triplet lens type
  • 20x minimum magnification
  • 53/64 in (21.03 mm) lens diameter
  • Hand-held design
  • Features 3 precision ground lens elements
  • Foldable design
  • No light source included