Learn More
MSC SPI Magnetic Indicator Base, 176 lb Magnetic Pull, M8 Thread
Catalog No. 5000530157
Encompass
This magnetic indicator base provides firm adsorption and is non-destructive to the workpiece. It features a stem mount and a strong magnetic pull for secure positioning in various applications.
  • Magnetic base type.
  • Stem mount.
  • 176 lb magnetic pull.
  • Base dimensions are 60 mm length, 50 mm width, and 55 mm height.
  • Plain base surface.
  • M8 thread size.
  • Firm adsorption to surfaces.
  • Non-destructive to workpieces.
  • Not calibrated.
  • Traceability certification is not included.
Catalog No. 50-005-30157 Supplier MSC Supplier No. 99669897
This magnetic indicator base provides firm adsorption and is non-destructive to the workpiece. It features a stem mount and a strong magnetic pull for secure positioning in various applications.
  • Magnetic base type.
  • Stem mount.
  • 176 lb magnetic pull.
  • Base dimensions are 60 mm length, 50 mm width, and 55 mm height.
  • Plain base surface.
  • M8 thread size.
  • Firm adsorption to surfaces.
  • Non-destructive to workpieces.
  • Not calibrated.
  • Traceability certification is not included.