Learn More
MSC SPI Mechanical Depth Micrometer, 0-6 in Range, 0.001 in Graduation, 2.5 in Base, 6 Rods
Catalog No. 5000528549
Encompass
The SPI Mechanical Depth Micrometer is a precision instrument designed for accurate depth measurements. It offers a 0-6 inch measuring range with 0.001 inch graduations and features a 2.5 inch base with six interchangeable rods. This tool is ideal for professionals requiring reliable and repeatable depth readings in various applications.
  • Ratchet stop thimble for consistent measuring force.
  • Positive locking clamp ensures stable readings.
  • Includes a short form certificate of calibration.
  • Furnished in a fitted case for protection and storage.
  • Torque wrench included for convenience.
  • Accuracy of +/- 0.0002 "
  • NIST certified traceability.
Catalog No. 50-005-28549 Supplier MSC Supplier No. 42416362
The SPI Mechanical Depth Micrometer is a precision instrument designed for accurate depth measurements. It offers a 0-6 inch measuring range with 0.001 inch graduations and features a 2.5 inch base with six interchangeable rods. This tool is ideal for professionals requiring reliable and repeatable depth readings in various applications.
  • Ratchet stop thimble for consistent measuring force.
  • Positive locking clamp ensures stable readings.
  • Includes a short form certificate of calibration.
  • Furnished in a fitted case for protection and storage.
  • Torque wrench included for convenience.
  • Accuracy of +/- 0.0002 "
  • NIST certified traceability.