Learn More
MSC SPI Mechanical Depth Micrometer, 4 in, 0.001 in, 2.5 in Base
Catalog No. 5000528379
Encompass
This mechanical depth micrometer is designed for precise depth measurements, featuring a 4 inch range and 0.001 inch graduation. It includes four rods and a 2.5 inch base, making it suitable for various applications requiring accurate depth readings.
  • Calibrated for accuracy
  • Accuracy of 0.0002 inch
  • Digital counter for easy reading
  • Ratchet stop thimble ensures consistent measurement force
  • Hardened steel measuring face material for durability
  • NIST certified traceability provides reliable measurements
  • Includes a protective case for storage and transport
Catalog No. 50-005-28379 Supplier MSC Supplier No. 38173316
This mechanical depth micrometer is designed for precise depth measurements, featuring a 4 inch range and 0.001 inch graduation. It includes four rods and a 2.5 inch base, making it suitable for various applications requiring accurate depth readings.
  • Calibrated for accuracy
  • Accuracy of 0.0002 inch
  • Digital counter for easy reading
  • Ratchet stop thimble ensures consistent measurement force
  • Hardened steel measuring face material for durability
  • NIST certified traceability provides reliable measurements
  • Includes a protective case for storage and transport