Learn More
MSC SPI Mechanical Depth Micrometer, 6 in Range, 0.001 in Graduation, 4 in Base, 6 Rods
Catalog No. 5000528378
Encompass
This SPI Mechanical Depth Micrometer is designed for precise depth measurements, featuring a 6-inch range and 0.001-inch graduation. It comes with a 4-inch base and 6 interchangeable rods, making it suitable for various applications requiring accurate depth readings.
  • Ratchet stop thimble ensures consistent measuring force.
  • Rotating rod type for versatile measurement.
  • Hardened steel measuring face material provides durability.
  • Calibrated for accuracy.
  • Accuracy of 0.0002 "
  • Digital counter for easy reading.
  • Includes a short form certificate of calibration and a protective case.
  • NIST certified traceability for reliable standards.
  • NIST certification included.
  • Six measuring rods for extended range.
Catalog No. 50-005-28378 Supplier MSC Supplier No. 38173290
This SPI Mechanical Depth Micrometer is designed for precise depth measurements, featuring a 6-inch range and 0.001-inch graduation. It comes with a 4-inch base and 6 interchangeable rods, making it suitable for various applications requiring accurate depth readings.
  • Ratchet stop thimble ensures consistent measuring force.
  • Rotating rod type for versatile measurement.
  • Hardened steel measuring face material provides durability.
  • Calibrated for accuracy.
  • Accuracy of 0.0002 "
  • Digital counter for easy reading.
  • Includes a short form certificate of calibration and a protective case.
  • NIST certified traceability for reliable standards.
  • NIST certification included.
  • Six measuring rods for extended range.