Learn More
MSC SPI Mechanical Hole Micrometer, 0.2400-0.2800 in, 0.0001 in Accuracy
Catalog No. 5000533748
Encompass
This mechanical hole micrometer provides accurate measurement of internal diameters. It is equipped with self-centering contact points and a ratchet stop, ensuring consistent and precise readings, even for measurements near the bottom of blind holes.
  • Self-centering contact points for accurate internal diameter measurements
  • Measuring contact points allow measurements near the bottom of blind holes
  • Supplied with calibrated setting ring
  • Designed to accommodate additional extensions for increased measuring depth
  • Ratchet stop for consistent readings
  • Features 2 measuring points
  • Includes a short form calibration certificate
  • Furnished in a fitted case
  • Two year limited warranty
  • Ratchet thimble for precise control
  • Carbide measuring surfaces for durability
Catalog No. 50-005-33748 Supplier MSC Supplier No. 37881117
This mechanical hole micrometer provides accurate measurement of internal diameters. It is equipped with self-centering contact points and a ratchet stop, ensuring consistent and precise readings, even for measurements near the bottom of blind holes.
  • Self-centering contact points for accurate internal diameter measurements
  • Measuring contact points allow measurements near the bottom of blind holes
  • Supplied with calibrated setting ring
  • Designed to accommodate additional extensions for increased measuring depth
  • Ratchet stop for consistent readings
  • Features 2 measuring points
  • Includes a short form calibration certificate
  • Furnished in a fitted case
  • Two year limited warranty
  • Ratchet thimble for precise control
  • Carbide measuring surfaces for durability