Learn More
MSC SPI Mechanical Hole Micrometer, 0.6500 to 0.8000 in Range, 3-Point
Catalog No. 5000527944
Encompass
This mechanical hole micrometer measures internal diameters from 0.6500 to 0.8000 inches with 0.0002 inch graduation and 0.0001 inch accuracy. It features a ratchet stop thimble for consistent measuring force and self-centering contact points for high accuracy, even near blind hole bottoms. A calibrated setting ring and optional extensions enhance versatility.
  • Self-centering contact points for accurate internal diameter measurement.
  • Measuring contact points designed for measurements near blind hole bottoms.
  • Supplied with a calibrated setting ring.
  • Additional extensions can be added to increase the measuring depth.
  • Ratchet stop.
  • Short form calibration certificate included.
  • Furnished in a fitted case.
  • Carbide measuring surfaces.
  • Two-year limited warranty.
Catalog No. 50-005-27944 Supplier MSC Supplier No. 37881059
This mechanical hole micrometer measures internal diameters from 0.6500 to 0.8000 inches with 0.0002 inch graduation and 0.0001 inch accuracy. It features a ratchet stop thimble for consistent measuring force and self-centering contact points for high accuracy, even near blind hole bottoms. A calibrated setting ring and optional extensions enhance versatility.
  • Self-centering contact points for accurate internal diameter measurement.
  • Measuring contact points designed for measurements near blind hole bottoms.
  • Supplied with a calibrated setting ring.
  • Additional extensions can be added to increase the measuring depth.
  • Ratchet stop.
  • Short form calibration certificate included.
  • Furnished in a fitted case.
  • Carbide measuring surfaces.
  • Two-year limited warranty.