Learn More
MSC SPI Mechanical Hole Micrometer, 1.6000 to 2.0000 in Range, 0.0002 Accuracy, 3-Point
Catalog No. 5000533677
Encompass
The SPI Mechanical Hole Micrometer is engineered for precise internal diameter measurements. It features self-centering contact points and durable carbide measuring surfaces to ensure accuracy and consistency. Equipped with a ratchet stop, this micrometer provides reliable readings for various industrial and laboratory applications.
  • Self-centering contact points for accurate measurement of internal diameters.
  • Measuring contact points allow measurements near the bottom of blind holes.
  • Supplied with a calibrated setting ring.
  • Additional extensions can be added to increase measuring depth.
  • Includes a ratchet stop.
  • Carbide measuring surfaces.
  • Furnished in a fitted case.
  • Comes with a two year limited warranty.
  • Short form calibration certificate included.
Catalog No. 50-005-33677 Supplier MSC Supplier No. 37881000
The SPI Mechanical Hole Micrometer is engineered for precise internal diameter measurements. It features self-centering contact points and durable carbide measuring surfaces to ensure accuracy and consistency. Equipped with a ratchet stop, this micrometer provides reliable readings for various industrial and laboratory applications.
  • Self-centering contact points for accurate measurement of internal diameters.
  • Measuring contact points allow measurements near the bottom of blind holes.
  • Supplied with a calibrated setting ring.
  • Additional extensions can be added to increase measuring depth.
  • Includes a ratchet stop.
  • Carbide measuring surfaces.
  • Furnished in a fitted case.
  • Comes with a two year limited warranty.
  • Short form calibration certificate included.