Learn More
MSC SPI Mechanical Hole Micrometer, 6 to 7 in. Range, 0.0002 in. Graduation, 0.000400 in. Accuracy
Catalog No. 5000534139
Encompass
This mechanical hole micrometer is designed for precise internal measurements with a range of 6 to 7 in. It offers high accuracy with a 0.0002 in. graduation and 0.000400 in. accuracy, featuring a ratchet stop thimble for consistent pressure.
  • Graduation of 0.0002 in.
  • Accuracy of 0.000400 in.
  • Ratchet stop thimble for consistent measurements.
  • Self aligning & self centering mechanism.
  • Carbide measuring anvils for durability.
Catalog No. 50-005-34139 Supplier MSC Supplier No. 00343103
This mechanical hole micrometer is designed for precise internal measurements with a range of 6 to 7 in. It offers high accuracy with a 0.0002 in. graduation and 0.000400 in. accuracy, featuring a ratchet stop thimble for consistent pressure.
  • Graduation of 0.0002 in.
  • Accuracy of 0.000400 in.
  • Ratchet stop thimble for consistent measurements.
  • Self aligning & self centering mechanism.
  • Carbide measuring anvils for durability.