Learn More
MSC SPI Mechanical Inside Micrometer, 0.2000-0.2400 in, 0.0001 in Accuracy
Catalog No. 5000532377
Encompass
The SPI Mechanical Inside Micrometer is designed for precise internal diameter measurements. It features a range of 0.2000 to 0.2400 in with an accuracy of 0.0001 in. This tool is ideal for applications requiring accurate dimensional control in laboratory and industrial settings.
  • Self-centering contact points for accurate internal diameter measurement.
  • Measures near the bottom of blind holes.
  • Supplied with calibrated setting ring.
  • Additional extensions can increase measuring depth.
  • Ratchet stop thimble.
  • Includes 2 measuring points for models up to 0.24 in.
  • Short form calibration certificate included.
  • Furnished in fitted case.
  • Two year limited warranty.
Catalog No. 50-005-32377 Supplier MSC Supplier No. 37881125
The SPI Mechanical Inside Micrometer is designed for precise internal diameter measurements. It features a range of 0.2000 to 0.2400 in with an accuracy of 0.0001 in. This tool is ideal for applications requiring accurate dimensional control in laboratory and industrial settings.
  • Self-centering contact points for accurate internal diameter measurement.
  • Measures near the bottom of blind holes.
  • Supplied with calibrated setting ring.
  • Additional extensions can increase measuring depth.
  • Ratchet stop thimble.
  • Includes 2 measuring points for models up to 0.24 in.
  • Short form calibration certificate included.
  • Furnished in fitted case.
  • Two year limited warranty.