Learn More
MSC SPI Mechanical Inside Micrometer, 0.2000-1.2000 in Range, 0.001 in Graduation
Catalog No. 5000528668
Encompass
This mechanical inside micrometer provides precise internal measurements for various applications. It offers a measuring range of 0.2000 to 1.2000 inches with a fine graduation of 0.001 inch, ensuring accuracy and reliability for detailed work.
  • Ratchet stop thimble for consistent measuring force
  • Calibrated for accuracy
  • Satin chrome finish for durability
  • NIST traceability certification included
  • Single contact point
  • Includes one setting ring
  • Two-year warranty
  • Carbide measuring faces for extended life
  • Short form certificate of calibration included
Catalog No. 50-005-28668 Supplier MSC Supplier No. 38169108
This mechanical inside micrometer provides precise internal measurements for various applications. It offers a measuring range of 0.2000 to 1.2000 inches with a fine graduation of 0.001 inch, ensuring accuracy and reliability for detailed work.
  • Ratchet stop thimble for consistent measuring force
  • Calibrated for accuracy
  • Satin chrome finish for durability
  • NIST traceability certification included
  • Single contact point
  • Includes one setting ring
  • Two-year warranty
  • Carbide measuring faces for extended life
  • Short form certificate of calibration included