Learn More
MSC SPI Mechanical Inside Micrometer, 1.6000 to 4.0000 in., Carbide Anvils
Catalog No. 5000534134
Encompass
This mechanical inside micrometer set provides precise measurement of bore diameters with self-centering and self-aligning features. Equipped with durable carbide measuring anvils and a ratchet stop, it ensures high accuracy and repeatability. Ideal for diverse laboratory and industrial applications, this set measures a range of 1.60 to 4.00 inches.
  • Self-centering and self-aligning design
  • Carbide tipped measuring anvils
  • Ratchet stop for accuracy and repeatability
  • Three-point internal micrometers for bore diameters
  • Measures within .050″ of blind hole bottom (up to 4″ capacity)
  • Tapered feelers for positive reading of the front edge
  • Includes extension rod, setting rings, and spanner wrenches in a wooden case
  • NIST certification available upon request
Catalog No. 50-005-34134 Supplier MSC Supplier No. 00343095
This mechanical inside micrometer set provides precise measurement of bore diameters with self-centering and self-aligning features. Equipped with durable carbide measuring anvils and a ratchet stop, it ensures high accuracy and repeatability. Ideal for diverse laboratory and industrial applications, this set measures a range of 1.60 to 4.00 inches.
  • Self-centering and self-aligning design
  • Carbide tipped measuring anvils
  • Ratchet stop for accuracy and repeatability
  • Three-point internal micrometers for bore diameters
  • Measures within .050″ of blind hole bottom (up to 4″ capacity)
  • Tapered feelers for positive reading of the front edge
  • Includes extension rod, setting rings, and spanner wrenches in a wooden case
  • NIST certification available upon request