Learn More
MSC SPI Mechanical Inside Micrometer, 5.00 to 30.00 mm, 0.01 mm Graduation, Ratchet Stop
Catalog No. 5000528345
Encompass
This mechanical inside micrometer is designed for precise internal measurements within a range of 5.00 to 30.00 mm. It offers a graduation of 0.01 mm and features a ratchet stop thimble to ensure consistent measuring force. Durable carbide measuring faces contribute to long-term accuracy, and it includes a setting ring and a certificate of calibration for immediate use.
  • Measures from 5.00 to 30.00 mm
  • 0.01 mm graduation for precise readings
  • Ratchet stop thimble ensures consistent measuring force
  • Carbide measuring faces provide durability
  • Includes a setting ring
  • Supplied with a short form certificate of calibration
  • Backed by a 2 year warranty
Catalog No. 50-005-28345 Supplier MSC Supplier No. 38169074
This mechanical inside micrometer is designed for precise internal measurements within a range of 5.00 to 30.00 mm. It offers a graduation of 0.01 mm and features a ratchet stop thimble to ensure consistent measuring force. Durable carbide measuring faces contribute to long-term accuracy, and it includes a setting ring and a certificate of calibration for immediate use.
  • Measures from 5.00 to 30.00 mm
  • 0.01 mm graduation for precise readings
  • Ratchet stop thimble ensures consistent measuring force
  • Carbide measuring faces provide durability
  • Includes a setting ring
  • Supplied with a short form certificate of calibration
  • Backed by a 2 year warranty