Learn More
MSC SPI Mechanical Inside Micrometer, 60-70 mm Range, 0.001 in Graduation, 0.002 mm Accuracy
Catalog No. 5000536775
Encompass
This mechanical inside micrometer offers precise internal measurements with a range of 60 to 70 mm. It provides a graduation of 0.001 inch and an accuracy of 0.002 mm, featuring a ratchet stop thimble for consistent pressure and solid carbide measuring faces for durability.
  • Measures internal dimensions from 60 to 70 mm
  • Provides accurate readings with 0.001 inch graduation
  • Ensures precise measurements with 0.002 mm accuracy
  • Features a ratchet stop thimble for consistent measuring force
  • Equipped with solid carbide measuring faces for extended tool life
Catalog No. 50-005-36775 Supplier MSC Supplier No. 85199552
This mechanical inside micrometer offers precise internal measurements with a range of 60 to 70 mm. It provides a graduation of 0.001 inch and an accuracy of 0.002 mm, featuring a ratchet stop thimble for consistent pressure and solid carbide measuring faces for durability.
  • Measures internal dimensions from 60 to 70 mm
  • Provides accurate readings with 0.001 inch graduation
  • Ensures precise measurements with 0.002 mm accuracy
  • Features a ratchet stop thimble for consistent measuring force
  • Equipped with solid carbide measuring faces for extended tool life