Learn More
MSC SPI Mechanical Inside Micrometer Set, 0.1500-0.2750 in, Ratchet Stop, Carbide Anvils
Catalog No. 5000513289
Encompass
This mechanical inside micrometer set is designed for precise internal measurements within a range of 0.1500 to 0.2750 " It features self-aligning and self-centering capabilities, ensuring accurate readings. The set includes a ratchet stop thimble and durable carbide measuring anvils for consistent performance and longevity.
  • Measures from 0.1500 to 0.2750 inch
  • Self-aligning and self-centering
  • Carbide measuring anvils
  • Ratchet stop thimble
  • Accuracy of 0.000200 inch
  • Includes setting rings and spanner wrenches in a wooden case
  • Three measuring heads
Catalog No. 50-005-13289 Supplier MSC Supplier No. 00343194
This mechanical inside micrometer set is designed for precise internal measurements within a range of 0.1500 to 0.2750 " It features self-aligning and self-centering capabilities, ensuring accurate readings. The set includes a ratchet stop thimble and durable carbide measuring anvils for consistent performance and longevity.
  • Measures from 0.1500 to 0.2750 inch
  • Self-aligning and self-centering
  • Carbide measuring anvils
  • Ratchet stop thimble
  • Accuracy of 0.000200 inch
  • Includes setting rings and spanner wrenches in a wooden case
  • Three measuring heads