Learn More
MSC SPI Mechanical Micrometer Head, 0-0.5 in Range, 0.0002 in Accuracy
Catalog No. 5000536589
Encompass
This mechanical micrometer head is designed for improving accuracy in various applications such as machines, gauging, test equipment, electronics, optical instruments, and machine instruments. It features a flat spindle and includes a short form certificate of calibration.
  • Accurate up to 0.0002 in
  • Flat spindle
  • Calibrated and includes a short form certificate of calibration
  • Features a satin chrome finish
  • Clamp nut supplied
  • Improves accuracy in machines, gauging, and test equipment
  • Ideal for electronics, optical, and machine instruments
  • 2.3 in length
Catalog No. 50-005-36589 Supplier MSC Supplier No. 76833490
This mechanical micrometer head is designed for improving accuracy in various applications such as machines, gauging, test equipment, electronics, optical instruments, and machine instruments. It features a flat spindle and includes a short form certificate of calibration.
  • Accurate up to 0.0002 in
  • Flat spindle
  • Calibrated and includes a short form certificate of calibration
  • Features a satin chrome finish
  • Clamp nut supplied
  • Improves accuracy in machines, gauging, and test equipment
  • Ideal for electronics, optical, and machine instruments
  • 2.3 in length