Learn More
MSC SPI Mechanical Outside Micrometer, 5 in Range, 0.0001 in Graduation, Ratchet Stop Thimble
Catalog No. 5000527744
Encompass
This mechanical outside micrometer features a 5 inch range with 0.0001 inch graduation and 0.0002 inch accuracy. It includes a ratchet stop thimble, rotating spindle, and is calibrated for precision measurements.
  • Mechanical digit counter for quick, error-free reading
  • Carbide measuring face
  • Standards included above 0-1 inch range/0-25 mm
  • Ratchet thimble
  • Spindle lock
  • Short form calibration certificate included
  • Furnished in fitted case
  • Two year limited warranty
Catalog No. 50-005-27744 Supplier MSC Supplier No. 38173175
This mechanical outside micrometer features a 5 inch range with 0.0001 inch graduation and 0.0002 inch accuracy. It includes a ratchet stop thimble, rotating spindle, and is calibrated for precision measurements.
  • Mechanical digit counter for quick, error-free reading
  • Carbide measuring face
  • Standards included above 0-1 inch range/0-25 mm
  • Ratchet thimble
  • Spindle lock
  • Short form calibration certificate included
  • Furnished in fitted case
  • Two year limited warranty