Learn More
MSC SPI Threaded Ring Gage, M10 x 1.50, Class 6G, No Go, 1 ea
Catalog No. 5000514401
Encompass
This metric threaded ring gage is designed for precision inspection of external threads. Constructed from durable tool steel, this No Go gage ensures components meet the required specifications for M10 x 1.50 threads with a 6G classification.
  • Features M10 x 1.50 thread size
  • Class 6G classification for tight tolerance inspection
  • Manufactured from wear-resistant tool steel
  • Includes NPL traceability certification
Catalog No. 50-005-14401 Supplier MSC Supplier No. 75890434
This metric threaded ring gage is designed for precision inspection of external threads. Constructed from durable tool steel, this No Go gage ensures components meet the required specifications for M10 x 1.50 threads with a 6G classification.
  • Features M10 x 1.50 thread size
  • Class 6G classification for tight tolerance inspection
  • Manufactured from wear-resistant tool steel
  • Includes NPL traceability certification