Learn More
MSC SPI Threaded Ring Gage, M4.5 x 0.75, 6G, No Go, 1 ea
Catalog No. 5000512657
Encompass
This metric threaded ring gage is designed for precise measurement and inspection of M4.5 x 0.75 threads. As a No Go gage, it is essential for verifying that internal threads do not exceed specified tolerance limits during production or quality control processes. The gage is manufactured in accordance with ISO 1502 standards to ensure accuracy and consistency.
  • Metric thread type with M4.5 x 0.75 size
  • Class 6G classification
  • Single ring design for No Go inspection
  • Constructed from durable tool steel
  • Supplied with a certificate of calibration traceable to NPL standards
Catalog No. 50-005-12657 Supplier MSC Supplier No. 75890335
This metric threaded ring gage is designed for precise measurement and inspection of M4.5 x 0.75 threads. As a No Go gage, it is essential for verifying that internal threads do not exceed specified tolerance limits during production or quality control processes. The gage is manufactured in accordance with ISO 1502 standards to ensure accuracy and consistency.
  • Metric thread type with M4.5 x 0.75 size
  • Class 6G classification
  • Single ring design for No Go inspection
  • Constructed from durable tool steel
  • Supplied with a certificate of calibration traceable to NPL standards