Learn More
MSC SPI Threaded Ring Gage, M30 x 2.00, Class 6G, No Go, 1 ea
Catalog No. 5000511589
Encompass
This metric thread ring gage is designed for precision inspection of M30 x 2.00 threads. As a No Go gage, it is essential for verifying that thread dimensions remain within the specified 6G tolerance class, helping to ensure manufacturing quality and adherence to strict engineering standards.
  • Constructed from durable tool steel for long-lasting wear resistance
  • Manufactured with thread dimensions compliant with ISO 1502 standards
  • Includes a certificate of calibration traceable to the National Physical Laboratory (NPL)
  • Single-ended design for efficient use
Catalog No. 50-005-11589 Supplier MSC Supplier No. 75890855
This metric thread ring gage is designed for precision inspection of M30 x 2.00 threads. As a No Go gage, it is essential for verifying that thread dimensions remain within the specified 6G tolerance class, helping to ensure manufacturing quality and adherence to strict engineering standards.
  • Constructed from durable tool steel for long-lasting wear resistance
  • Manufactured with thread dimensions compliant with ISO 1502 standards
  • Includes a certificate of calibration traceable to the National Physical Laboratory (NPL)
  • Single-ended design for efficient use