Learn More
MSC SPI Micrometer Anvil, For 1 to 12 in Micrometer, Comparator
Catalog No. 5000511240
Encompass
The micrometer anvil is designed for use with micrometers ranging from 1 to 12 inches. It functions as a comparator anvil, providing a stable contact point for accurate measurements.
  • Designed for use with 1 to 12 inch micrometers.
  • Functions as a comparator anvil.
  • Provides a stable contact point for accurate measurements.
Catalog No. 50-005-11240 Supplier MSC Supplier No. 06404255
The micrometer anvil is designed for use with micrometers ranging from 1 to 12 inches. It functions as a comparator anvil, providing a stable contact point for accurate measurements.
  • Designed for use with 1 to 12 inch micrometers.
  • Functions as a comparator anvil.
  • Provides a stable contact point for accurate measurements.