Learn More
MSC SPI Micrometer Calibration Standard, 10 in, 0.0002 in Accuracy, Flat End
Catalog No. 5000513791
Encompass
This Micrometer Calibration Standard is designed for precise measurements and calibration of micrometers. It features a robust design and includes a heat insulating handle for user comfort and accuracy.
  • Maximum length is 10 in
  • Accuracy is 0.000240 in
  • Flat end shape
  • Includes heat insulating handle
  • Calibrated
  • For use with micrometers
Catalog No. 50-005-13791 Supplier MSC Supplier No. 06407407
This Micrometer Calibration Standard is designed for precise measurements and calibration of micrometers. It features a robust design and includes a heat insulating handle for user comfort and accuracy.
  • Maximum length is 10 in
  • Accuracy is 0.000240 in
  • Flat end shape
  • Includes heat insulating handle
  • Calibrated
  • For use with micrometers