Learn More
MSC SPI Micrometer Calibration Standard, 11 in Length, 0.000280 in Accuracy, Flat End
Catalog No. 5000512722
Encompass
This micrometer calibration standard features an 11 inch length and a flat end, designed for precise calibration of micrometers. It offers an accuracy of 0.000280 inch and includes a heat insulating handle for comfortable use.
  • Provides an 11 inch maximum length for calibration.
  • Achieves an accuracy of 0.000280 "
  • Features a flat end shape for stable measurements.
  • Comes calibrated for immediate use.
  • Includes a heat insulating handle for comfortable operation.
  • Suitable for calibrating micrometers.
Catalog No. 50-005-12722 Supplier MSC Supplier No. 06407415
This micrometer calibration standard features an 11 inch length and a flat end, designed for precise calibration of micrometers. It offers an accuracy of 0.000280 inch and includes a heat insulating handle for comfortable use.
  • Provides an 11 inch maximum length for calibration.
  • Achieves an accuracy of 0.000280 "
  • Features a flat end shape for stable measurements.
  • Comes calibrated for immediate use.
  • Includes a heat insulating handle for comfortable operation.
  • Suitable for calibrating micrometers.