Learn More
MSC SPI Micrometer Setting Rings, 4 to 5 in Range
Catalog No. 5000529965
Encompass
These Micrometer Setting Rings are designed for use with SPI Hole-Mike Series Micrometers. They are specifically used for precise measurements ranging from 4 to 5 inches.
  • Accessory type is setting ring
  • For use with Hole-Mike Series micrometers
  • Maximum measurement is 5.0000 in
  • Country of origin is India
Catalog No. 50-005-29965 Supplier MSC Supplier No. 00343012
These Micrometer Setting Rings are designed for use with SPI Hole-Mike Series Micrometers. They are specifically used for precise measurements ranging from 4 to 5 inches.
  • Accessory type is setting ring
  • For use with Hole-Mike Series micrometers
  • Maximum measurement is 5.0000 in
  • Country of origin is India