Learn More
MSC SPI Micrometer Setting Rings, 7 in., For SPI Hole-Mike Series Micrometer
Catalog No. 5000534240
Encompass
This setting ring is designed for use with Hole-Mike Series Micrometers. It provides a precise 7 in. measurement for accurate calibration.
  • Provides a 7 in. measurement
  • Designed for use with Hole-Mike Series Micrometers
  • Accessory type: setting ring
Catalog No. 50-005-34240 Supplier MSC Supplier No. 00343079
This setting ring is designed for use with Hole-Mike Series Micrometers. It provides a precise 7 in. measurement for accurate calibration.
  • Provides a 7 in. measurement
  • Designed for use with Hole-Mike Series Micrometers
  • Accessory type: setting ring