Learn More
MSC SPI Micrometer Spare Part Kit, for Outside Micrometers, 4 Pieces
Catalog No. 5000511437
Encompass
This micrometer spare part kit is designed for use with outside micrometers, specifically for 0-12 in IP54 (2 button) digital outside micrometers. It includes four essential pieces to maintain and repair your precision measurement equipment.
  • Spare part kit for micrometers
  • Designed for use with outside micrometers
  • Includes parts for 0-12 in IP54 digital outside micrometers
  • Kit contains a fixed stop, knurled thimble, locking device, and battery cover
  • Comprises 4 pieces
Catalog No. 50-005-11437 Supplier MSC Supplier No. 03699014
This micrometer spare part kit is designed for use with outside micrometers, specifically for 0-12 in IP54 (2 button) digital outside micrometers. It includes four essential pieces to maintain and repair your precision measurement equipment.
  • Spare part kit for micrometers
  • Designed for use with outside micrometers
  • Includes parts for 0-12 in IP54 digital outside micrometers
  • Kit contains a fixed stop, knurled thimble, locking device, and battery cover
  • Comprises 4 pieces