Learn More
MSC SPI Micrometer Thread Triangle, Any Micrometer Spindle, 4-80 Pitch, 2 Pieces
Catalog No. 5000533442
Encompass
The Micrometer Thread Triangle is designed for use with any micrometer spindle. This two-piece set includes holders and a chart, covering a thread pitch range of 4.00 to 80.00.
  • Thread triangle accessory
  • Compatible with any micrometer spindle
  • Two-piece set
  • Includes holders and chart
  • Covers a pitch range of 4.00 to 80.00
Catalog No. 50-005-33442 Supplier MSC Supplier No. 01718303
The Micrometer Thread Triangle is designed for use with any micrometer spindle. This two-piece set includes holders and a chart, covering a thread pitch range of 4.00 to 80.00.
  • Thread triangle accessory
  • Compatible with any micrometer spindle
  • Two-piece set
  • Includes holders and chart
  • Covers a pitch range of 4.00 to 80.00