Learn More
MSC SPI OD Feeler, 2 in Long, Co-Ax Centering Indicator
Catalog No. 5000529100
Encompass
This OD feeler is designed for use with Co-Ax Centering Indicators. It has a length of 2 inches, providing a simplified and precise method for centering workpieces or fixtures on horizontal or vertical machines. The design allows the indicator face to remain stationary, enabling hands-free operation for accurate machine table positioning.
  • Measures 2 in in length.
  • Designed for use with Co-Ax Centering Indicators.
  • Facilitates precise centering of workpieces or fixtures.
  • Allows indicator face to remain stationary during spindle rotation.
  • Enables hands-free machine table positioning.
  • Operating speed should not exceed 800 RPM.
  • Suitable for internal and external diameter measurements.
Catalog No. 50-005-29100 Supplier MSC Supplier No. 09232794
This OD feeler is designed for use with Co-Ax Centering Indicators. It has a length of 2 inches, providing a simplified and precise method for centering workpieces or fixtures on horizontal or vertical machines. The design allows the indicator face to remain stationary, enabling hands-free operation for accurate machine table positioning.
  • Measures 2 in in length.
  • Designed for use with Co-Ax Centering Indicators.
  • Facilitates precise centering of workpieces or fixtures.
  • Allows indicator face to remain stationary during spindle rotation.
  • Enables hands-free machine table positioning.
  • Operating speed should not exceed 800 RPM.
  • Suitable for internal and external diameter measurements.