Learn More
MSC SPI Optical Comparator Chart & Reticle, 1-3/8 in Diameter, 0.005 in Increment, 10x Magnification
Catalog No. 5000527476
Encompass
This optical comparator chart and reticle features a 1-3/8 inch diameter and is designed for precise measurements with a 0.005 inch increment. It is suitable for use with 10x magnification, offering accurate visual inspection in various applications.
  • Overall diameter of 1-3/8 in
  • Increments of 0.005 in for detailed measurements
  • Designed for use with 10x magnification systems
Catalog No. 50-005-27476 Supplier MSC Supplier No. 06554133
This optical comparator chart and reticle features a 1-3/8 inch diameter and is designed for precise measurements with a 0.005 inch increment. It is suitable for use with 10x magnification, offering accurate visual inspection in various applications.
  • Overall diameter of 1-3/8 in
  • Increments of 0.005 in for detailed measurements
  • Designed for use with 10x magnification systems