Learn More
MSC SPI Optical Comparator Chart & Reticle, 7x Magnification, 1 in Diameter, Reticle #12
Catalog No. 5000513548
Encompass
This optical comparator chart and reticle is designed for precise measurement and inspection applications. It features a 1 inch overall diameter and is compatible with 7x magnification comparators.
  • For use with 7x magnification
  • 1 inch overall diameter
  • Reticle number 12
  • Compatible with mini comparators
  • Manufactured in Japan
Catalog No. 50-005-13548 Supplier MSC Supplier No. 06555122
This optical comparator chart and reticle is designed for precise measurement and inspection applications. It features a 1 inch overall diameter and is compatible with 7x magnification comparators.
  • For use with 7x magnification
  • 1 inch overall diameter
  • Reticle number 12
  • Compatible with mini comparators
  • Manufactured in Japan