Learn More
MSC SPI Optical Comparator Reticle, 1-3/8 in Dia, 10x, #6
Catalog No. 5000529555
Encompass
This SPI optical comparator reticle is designed for precision measurements and inspection tasks. It features a 1-3/8 inch diameter and is specifically engineered for use with 10x optical comparators, providing clear and accurate visual comparison. The reticle is designated as number 6.
  • Designed for 10x magnification optical comparators
  • Features a 1-3/8 inch overall diameter
  • Reticle number 6 for specific applications
  • Manufactured in Japan
Catalog No. 50-005-29555 Supplier MSC Supplier No. 06554067
This SPI optical comparator reticle is designed for precision measurements and inspection tasks. It features a 1-3/8 inch diameter and is specifically engineered for use with 10x optical comparators, providing clear and accurate visual comparison. The reticle is designated as number 6.
  • Designed for 10x magnification optical comparators
  • Features a 1-3/8 inch overall diameter
  • Reticle number 6 for specific applications
  • Manufactured in Japan