Learn More
MSC SPI Optical Comparator Reticle, 1 inch Diameter, No. 10, 7x Magnification
Catalog No. 5000528481
Encompass
This optical comparator reticle is designed for precision measurement and inspection tasks. It features a 1 inch diameter and is specified as Reticle Number 10, compatible with 7x magnification optical comparators.
  • Designed for use with 7x magnification optical comparators.
  • 1 inch overall diameter.
  • Reticle number 10.
  • Suitable for precision measurement and inspection.
Catalog No. 50-005-28481 Supplier MSC Supplier No. 06555106
This optical comparator reticle is designed for precision measurement and inspection tasks. It features a 1 inch diameter and is specified as Reticle Number 10, compatible with 7x magnification optical comparators.
  • Designed for use with 7x magnification optical comparators.
  • 1 inch overall diameter.
  • Reticle number 10.
  • Suitable for precision measurement and inspection.