Learn More
MSC SPI Oxide Pin Gage, Minus, Black, 0.799 in
Catalog No. 5000530199
Encompass
This oxide pin gage is colored black and has a size of 0.799 " It features a black oxide coating that prevents corrosion and indicates gage wear. The gage is made from hardened drill rod with a Rockwell C hardness of 60-62.
  • Black oxide coating prevents corrosion and shows gage wear.
  • Made from hardened drill rod RC 60-62.
  • 0.799 inch pin size.
  • 2 inch length.
  • Black color.
Catalog No. 50-005-30199 Supplier MSC Supplier No. 56251499
This oxide pin gage is colored black and has a size of 0.799 " It features a black oxide coating that prevents corrosion and indicates gage wear. The gage is made from hardened drill rod with a Rockwell C hardness of 60-62.
  • Black oxide coating prevents corrosion and shows gage wear.
  • Made from hardened drill rod RC 60-62.
  • 0.799 inch pin size.
  • 2 inch length.
  • Black color.