Learn More
MSC SPI Pin Gage, 0.1190 inch Diameter, Class ZZ Minus, Black Oxide
Catalog No. 5000514523
Encompass
This gage is a precision measuring tool designed for checking hole sizes, distances between holes, or slot widths. Featuring a 0.1190 inch diameter and Class ZZ Minus tolerance, it ensures accurate measurements required for quality control and manufacturing processes. The black oxide coating provides added durability and corrosion resistance, extending the life of the tool.
  • Ensures precise measurement of 0.1190 inch diameter.
  • Features class ZZ minus tolerance for high accuracy.
  • Includes a durable black oxide coating for corrosion resistance.
Catalog No. 50-005-14523 Supplier MSC Supplier No. 56244676
This gage is a precision measuring tool designed for checking hole sizes, distances between holes, or slot widths. Featuring a 0.1190 inch diameter and Class ZZ Minus tolerance, it ensures accurate measurements required for quality control and manufacturing processes. The black oxide coating provides added durability and corrosion resistance, extending the life of the tool.
  • Ensures precise measurement of 0.1190 inch diameter.
  • Features class ZZ minus tolerance for high accuracy.
  • Includes a durable black oxide coating for corrosion resistance.