Learn More
MSC SPI Pin Gage, 0.220 in, Black Oxide Finish, Class ZZ Plus
Catalog No. 5000531303
Encompass
This pin gage is designed for precise measurement and inspection. It features a black oxide coating for corrosion resistance and wear indication, and is made from hardened drill rod for durability.
  • Black oxide coating prevents corrosion and shows gage wear.
  • Made from hardened drill rod.
  • Overall length is 2 inches.
Catalog No. 50-005-31303 Supplier MSC Supplier No. 84500594
This pin gage is designed for precise measurement and inspection. It features a black oxide coating for corrosion resistance and wear indication, and is made from hardened drill rod for durability.
  • Black oxide coating prevents corrosion and shows gage wear.
  • Made from hardened drill rod.
  • Overall length is 2 inches.