Learn More
MSC SPI Pin Gage, 0.236 in, RC 60-62, Black Oxide
Catalog No. 5000515979
Encompass
This pin gage is designed for precise measurement and inspection tasks. It features a 0.236 inch diameter and is constructed from hardened drill rod with a Rockwell C hardness of 60-62. The gage is coated with black oxide, which not only prevents corrosion but also helps to show wear over time, indicating when replacement might be necessary. It has an overall length of 2 inches.
  • Black oxide coating prevents corrosion and shows gage wear.
  • Hardened drill rod with a hardness of RC 60-62.
  • Overall length of 2 inches.
  • Diameter of 0.236 inches.
Catalog No. 50-005-15979 Supplier MSC Supplier No. 84500750
This pin gage is designed for precise measurement and inspection tasks. It features a 0.236 inch diameter and is constructed from hardened drill rod with a Rockwell C hardness of 60-62. The gage is coated with black oxide, which not only prevents corrosion but also helps to show wear over time, indicating when replacement might be necessary. It has an overall length of 2 inches.
  • Black oxide coating prevents corrosion and shows gage wear.
  • Hardened drill rod with a hardness of RC 60-62.
  • Overall length of 2 inches.
  • Diameter of 0.236 inches.