Learn More
MSC SPI Pin Gage, 0.929 in Diameter, Class ZZ Minus, Black Oxide
Catalog No. 5000527990
Encompass
This pin gage is crafted from hardened drill rod with an RC 60-62 hardness, ensuring durability and precision for measurement applications. Its black oxide coating provides excellent corrosion resistance and clearly indicates gage wear, extending the product's lifespan.
  • Black oxide coating prevents corrosion and shows gage wear.
  • Hardened drill rod RC 60-62.
Catalog No. 50-005-27990 Supplier MSC Supplier No. 56252802
This pin gage is crafted from hardened drill rod with an RC 60-62 hardness, ensuring durability and precision for measurement applications. Its black oxide coating provides excellent corrosion resistance and clearly indicates gage wear, extending the product's lifespan.
  • Black oxide coating prevents corrosion and shows gage wear.
  • Hardened drill rod RC 60-62.