Learn More
MSC SPI Pin Gage, Black Oxide, 0.848 Inch, Plus
Catalog No. 5000528847
Encompass
This SPI Plus Pin Gage, sized at 0.848 inches, is designed for precise measurement and inspection tasks. It features a durable black oxide coating that helps prevent corrosion and serves as a visual indicator of wear, ensuring consistent accuracy over time. Constructed from hardened drill rod, this pin gage offers exceptional durability and reliability in various industrial applications.
  • Black oxide coating to prevent corrosion
  • Black oxide coating indicates gage wear
  • Made from hardened drill rod
  • RC hardness of 60-62
  • Overall length of 2 inches
  • Designed for precise measurement and inspection
Catalog No. 50-005-28847 Supplier MSC Supplier No. 56242035
This SPI Plus Pin Gage, sized at 0.848 inches, is designed for precise measurement and inspection tasks. It features a durable black oxide coating that helps prevent corrosion and serves as a visual indicator of wear, ensuring consistent accuracy over time. Constructed from hardened drill rod, this pin gage offers exceptional durability and reliability in various industrial applications.
  • Black oxide coating to prevent corrosion
  • Black oxide coating indicates gage wear
  • Made from hardened drill rod
  • RC hardness of 60-62
  • Overall length of 2 inches
  • Designed for precise measurement and inspection