Learn More
MSC SPI Pin Gage, Black Oxide, Class ZZ, Steel
Catalog No. 5000511747
Encompass
SPI Black Oxide Pin Gages are precision measuring tools designed for accurate diameter verification. Featuring a durable black oxide coating, these gages offer enhanced corrosion resistance and clearly show wear, ensuring reliable performance in demanding environments.
  • Prevents corrosion and shows gage wear
  • Hardened drill rod RC 60-62
  • Overall length 2 in
Catalog No. 50-005-11747 Supplier MSC Supplier No. 56252927
SPI Black Oxide Pin Gages are precision measuring tools designed for accurate diameter verification. Featuring a durable black oxide coating, these gages offer enhanced corrosion resistance and clearly show wear, ensuring reliable performance in demanding environments.
  • Prevents corrosion and shows gage wear
  • Hardened drill rod RC 60-62
  • Overall length 2 in